Assay Details
Target Gene Details
Entrez Gene ID: | 9261 |
Gene Name: | mitogen-activated protein kinase-activated protein kinase 2 |
Gene Aliases: |
MAPKAP-K2, MK-2, MK2 |
Location: |
Chr.1:206684912-206734285 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPKAPK2 | NM_004759.4 | NP_004750.1 | ||
| NM_032960.3 | NP_116584.2 | |||
| XM_005273353.3 | XP_005273410.1 | |||
| XM_017002810.1 | XP_016858299.1 | |||
| BC036060.1 | AAH36060.2 | |||
| BC052584.1 | AAH52584.1 | |||
| X75346.1 | CAA53094.1 |
More Information
Set Membership: |
|

Genomic Map