Assay Details
Target Gene Details
Entrez Gene ID: | 64129 |
Gene Name: | tubulointerstitial nephritis antigen like 1 |
Gene Aliases: |
ARG1, LCN7, LIECG3, TINAGRP |
Location: |
Chr.1:31576384-31587686 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TINAGL1 | XM_011541946.1 | XP_011540248.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011229 | Chr.1:31566071 - 31606805 on Build GRCh38 | Loss |
|
| nsv10062 | Chr.1:31571300 - 31623381 on Build GRCh38 | Gain+Loss |
|
| nsv950384 | Chr.1:31571200 - 31588799 on Build GRCh38 | Deletion |
|
| esv2758930 | Chr.1:31209533 - 32000730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map