Assay Details
Target Gene Details
Entrez Gene ID: | 51232 |
Gene Name: | cysteine rich transmembrane BMP regulator 1 |
Gene Aliases: |
CRIM-1, S52 |
Location: |
Chr.2:36356088-36551135 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRIM1 | NM_016441.2 | NP_057525.1 | ||
| XM_005264357.4 | XP_005264414.1 | |||
| XM_011532898.2 | XP_011531200.1 | |||
| XM_011532899.2 | XP_011531201.1 | |||
| XM_011532901.2 | XP_011531203.1 | |||
| XM_017004258.1 | XP_016859747.1 | |||
| XM_017004259.1 | XP_016859748.1 | |||
| XM_017004260.1 | XP_016859749.1 | |||
| XM_017004261.1 | XP_016859750.1 | |||
| XM_017004262.1 | XP_016859751.1 | |||
| AB209139.1 | BAD92376.1 | |||
| AF167706.1 | AAF34409.1 | |||
| AY358371.1 | AAQ88737.1 | |||
| BC111989.1 | AAI11990.1 | |||
| BC113371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2680 | Chr.2:36337775 - 36368588 on Build GRCh38 | Insertion |
|
| nsv833803 | Chr.2:36361922 - 36518636 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map