Assay Details
Target Gene Details
Entrez Gene ID: | 3032 |
Gene Name: | hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit |
Gene Aliases: |
ECHB, MSTP029, MTPB, TP-BETA |
Location: |
Chr.2:26244748-26290465 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HADHB | NM_000183.2 | NP_000174.1 | ||
| NM_001281512.1 | NP_001268441.1 | |||
| NM_001281513.1 | NP_001268442.1 | |||
| XM_011532803.1 | XP_011531105.1 | |||
| XM_017003939.1 | XP_016859428.1 | |||
| AF113209.1 | AAG39280.1 | |||
| AK130072.1 | ||||
| AK293142.1 | ||||
| AK302322.1 | ||||
| AK304455.1 | ||||
| AK314455.1 | ||||
| BC014572.1 | AAH14572.1 | |||
| BC017564.2 | AAH17564.1 | |||
| BC030824.1 | AAH30824.1 | |||
| BC066963.1 | AAH66963.1 | |||
| D16481.1 | BAA03942.1 | |||
| DA067388.1 | ||||
| DA835045.1 | ||||
| DB477128.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143044370] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map