Assay Details
Target Gene Details
| Entrez Gene ID: | 101928185 | 
| Gene Name: | uncharacterized LOC101928185 | 
| Gene Aliases: | - | 
| Location: | Chr.2:132915557-132931494 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| LOC101928185 | NR_135572.1 | |||
| DB058825.1 | 
Target Gene Details
| Entrez Gene ID: | 344148 | 
| Gene Name: | NCK associated protein 5 | 
| Gene Aliases: | ERIH1, ERIH2, NAP5 | 
| Location: | Chr.2:132671788-133675181 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 18 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1004604 | Chr.2:132900543 - 133004221 on Build GRCh38 | Loss |  NCKAP5  LOC101928185 | 
| nsv583158 | Chr.2:132558519 - 133104527 on Build GRCh38 | Gain |  NCKAP5  GPR39  LOC101928185  LYPD1 | 
| nsv583152 | Chr.2:132369502 - 133631550 on Build GRCh38 | Gain |  NCKAP5  GPR39  LOC101928161  LOC101928185  LYPD1 | 
| esv3592515 | Chr.2:132899712 - 132953048 on Build GRCh38 | Loss |  NCKAP5  LOC101928185 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top