Assay Details
Target Gene Details
Entrez Gene ID: | 130013 |
Gene Name: | aminocarboxymuconate semialdehyde decarboxylase |
Gene Aliases: |
- |
Location: |
Chr.2:134838319-134902906 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACMSD | XM_005263586.4 | 8 | 1866 | XP_005263643.1 |
| XM_005263588.4 | 9 | 1588 | XP_005263645.1 | |
| XM_005263589.4 | 11 | 2124 | XP_005263646.1 | |
| XM_005263590.4 | 7 | 2430 | XP_005263647.1 | |
| XM_011510592.2 | 10 | 2074 | XP_011508894.1 | |
| XM_017003325.1 | 10 | 2071 | XP_016858814.1 | |
| XM_017003326.1 | 10 | 2721 | XP_016858815.1 |
Target Gene Details
Entrez Gene ID: | 100129961 |
Gene Name: | CCNT2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:134866633-134918606 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNT2-AS1 | NR_036549.1 | |||
| AK303295.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75388224] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map