Assay Details
Target Gene Details
Entrez Gene ID: | 57683 |
Gene Name: | zinc finger DBF-type containing 2 |
Gene Aliases: |
- |
Location: |
Chr.2:206274641-206314438 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDBF2 | NM_001285549.1 | 7 | 8760 | NP_001272478.1 |
| NM_020923.2 | 5 | 8867 | NP_065974.1 | |
| XM_005246711.3 | 6 | 8867 | XP_005246768.1 | |
| XM_005246712.4 | 6 | 8690 | XP_005246769.1 | |
| XM_005246713.3 | 6 | 8684 | XP_005246770.1 | |
| XM_006712656.3 | 6 | 8723 | XP_006712719.1 | |
| XM_011511532.2 | 6 | 8900 | XP_011509834.1 | |
| XM_011511533.2 | 6 | 8717 | XP_011509835.1 | |
| XM_011511534.2 | 5 | 8640 | XP_011509836.1 | |
| XM_011511535.2 | 5 | 8646 | XP_011509837.1 | |
| XM_017004573.1 | 7 | 8973 | XP_016860062.1 | |
| XM_017004574.1 | 7 | 8899 | XP_016860063.1 | |
| XM_017004576.1 | 2 | 6869 | XP_016860065.1 | |
| AB046791.2 | 1 | 6735 | BAB13397.2 | |
| AB775780.1 | 7 | 8761 | ||
| AB775781.1 | 5 | 8867 | ||
| AK024339.1 | 1 | 703 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834516 | Chr.2:206282093 - 206440702 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116374070] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map