Assay Details
Target Gene Details
Entrez Gene ID: | 9759 |
Gene Name: | histone deacetylase 4 |
Gene Aliases: |
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA |
Location: |
Chr.2:239048168-239401647 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC4 | XM_011512221.1 | XP_011510523.1 | ||
| AK308791.1 |
Target Gene Details
Entrez Gene ID: | 101928111 |
Gene Name: | uncharacterized LOC101928111 |
Gene Aliases: |
- |
Location: |
Chr.2:239401436-239402364 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928111 | NR_135575.1 | |||
| BF434856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7357n54 | Chr.2:239400751 - 239401967 on Build GRCh38 | Loss |
|
| dgv689n67 | Chr.2:239392555 - 239406318 on Build GRCh38 | Gain |
|
| dgv7355n54 | Chr.2:239400274 - 239401967 on Build GRCh38 | Gain |
|
| nsv584781 | Chr.2:239397060 - 239401967 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map