Assay Details
Target Gene Details
Entrez Gene ID: | 57405 |
Gene Name: | SPC25, NDC80 kinetochore complex component |
Gene Aliases: |
AD024, SPBC25, hSpc25 |
Location: |
Chr.2:168865045-168890443 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPC25 | NM_020675.3 | NP_065726.1 | ||
| XM_011511516.2 | XP_011509818.1 | |||
| AF225416.1 | AAG09718.1 | |||
| AK291093.1 | ||||
| AY456388.1 | AAR88652.1 | |||
| BC022255.1 | AAH22255.1 | |||
| CR457197.1 | CAG33478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv227 | Chr.2:168864813 - 168882855 on Build GRCh38 | Insertion |
|
| nsv583599 | Chr.2:168807717 - 169046586 on Build GRCh38 | Gain |
|
| nsv3023 | Chr.2:168859032 - 168882855 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map