Assay Details
Target Gene Details
Entrez Gene ID: | 730100 |
Gene Name: | uncharacterized LOC730100 |
Gene Aliases: |
- |
Location: |
Chr.2:51032601-52407917 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC730100 | NR_135237.1 | |||
| AK127244.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6866n54 | Chr.2:50820175 - 51108720 on Build GRCh38 | Loss |
|
| nsv1000086 | Chr.2:50879876 - 51193400 on Build GRCh38 | Loss |
|
| nsv581848 | Chr.2:50711821 - 51118377 on Build GRCh38 | Loss |
|
| esv2763321 | Chr.2:50610356 - 51126248 on Build GRCh38 | Loss |
|
| nsv581853 | Chr.2:50786509 - 51326909 on Build GRCh38 | Loss |
|
| esv3892215 | Chr.2:50887566 - 51133529 on Build GRCh38 | Loss |
|
| nsv1002086 | Chr.2:50685023 - 51177923 on Build GRCh38 | Loss |
|
| nsv834014 | Chr.2:50954708 - 51133399 on Build GRCh38 | Loss |
|
| esv3892237 | Chr.2:51020156 - 51130764 on Build GRCh38 | Loss |
|
| nsv1012208 | Chr.2:51039660 - 51188856 on Build GRCh38 | Loss |
|

Genomic Map