Assay Details
Target Gene Details
Entrez Gene ID: | 646960 |
Gene Name: | protease, serine 56 |
Gene Aliases: |
MCOP6 |
Location: |
Chr.2:232520463-232525716 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS56 | NM_001195129.1 | NP_001182058.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527789 | Chr.2:232436106 - 232533505 on Build GRCh38 | Loss |
|
| nsv821918 | Chr.2:232319063 - 232560965 on Build GRCh38 | Gain |
|
| nsv1073020 | Chr.2:232525589 - 232526190 on Build GRCh38 | Deletion |
|
| nsv3211 | Chr.2:232515807 - 232527333 on Build GRCh38 | Insertion |
|
| nsv953202 | Chr.2:232474691 - 232546890 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map