Assay Details
Target Gene Details
Entrez Gene ID: | 84693 |
Gene Name: | methylmalonyl-CoA epimerase |
Gene Aliases: |
GLOD2 |
Location: |
Chr.2:71109676-71130288 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCEE | NM_032601.3 | NP_115990.3 | ||
| XM_005264613.2 | XP_005264670.1 | |||
| AB209469.1 | 1 | 528 | BAD92706.1 | |
| AF364547.1 | AAK52052.1 | |||
| AI033764.1 | ||||
| BC020825.1 | AAH20825.1 | |||
| BM271757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009014 | Chr.2:71096874 - 71181739 on Build GRCh38 | Gain |
|
| esv2662403 | Chr.2:71079295 - 71179547 on Build GRCh38 | Deletion |
|
| esv2759059 | Chr.2:71115012 - 71146290 on Build GRCh38 | Loss |
|
| nsv10002 | Chr.2:71114936 - 71121584 on Build GRCh38 | Loss |
|
| esv22790 | Chr.2:71115076 - 71120682 on Build GRCh38 | Loss |
|
| nsv517762 | Chr.2:71100222 - 71124357 on Build GRCh38 | Gain+Loss |
|
| nsv2771 | Chr.2:71108462 - 71145252 on Build GRCh38 | Deletion |
|
| esv2720185 | Chr.2:71079275 - 71185049 on Build GRCh38 | Deletion |
|
| nsv438353 | Chr.2:71105173 - 71115461 on Build GRCh38 | Loss |
|
| nsv834256 | Chr.2:71029484 - 71202682 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map