Assay Details
Target Gene Details
Entrez Gene ID: | 7535 |
Gene Name: | zeta chain of T cell receptor associated protein kinase 70 |
Gene Aliases: |
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70 |
Location: |
Chr.2:97712030-97744327 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZAP70 | NM_001079.3 | NP_001070.2 | ||
| XM_017004867.1 | XP_016860356.1 | |||
| XM_017004868.1 | XP_016860357.1 | |||
| XM_017004869.1 | XP_016860358.1 | |||
| XM_017004870.1 | XP_016860359.1 | |||
| AK303341.1 | ||||
| AK310628.1 | ||||
| AK310637.1 | ||||
| BC053878.1 | AAH53878.1 | |||
| L05148.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2837 | Chr.2:97694463 - 97728803 on Build GRCh38 | Insertion |
|
| esv33510 | Chr.2:97700640 - 97724839 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map