Assay Details
Target Gene Details
Entrez Gene ID: | 150678 |
Gene Name: | COP9 signalosome subunit 9 |
Gene Aliases: |
CSNAP, MYEOV2 |
Location: |
Chr.2:240126548-240136802 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPS9 | NM_001163424.1 | NP_001156896.1 | ||
| NM_138336.1 | NP_612209.1 | |||
| XM_017003411.1 | XP_016858900.1 | |||
| AF453951.1 | AAL41026.1 | |||
| AF487338.1 | AAL96264.2 | |||
| CN304277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997288 | Chr.2:239542278 - 240534009 on Build GRCh38 | Gain |
|

Genomic Map