Assay Details
Target Gene Details
Entrez Gene ID: | 285141 |
Gene Name: | glutamate rich 2 |
Gene Aliases: |
- |
Location: |
Chr.2:170770042-170798971 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERICH2 | NM_001290030.1 | NP_001276959.1 | ||
| NM_001290031.1 | NP_001276960.1 | |||
| NR_110644.1 | ||||
| XM_011510998.2 | XP_011509300.1 | |||
| XM_011510999.2 | XP_011509301.1 | |||
| XM_011511001.2 | XP_011509303.1 | |||
| XM_017003875.1 | XP_016859364.1 | |||
| XM_017003876.1 | XP_016859365.1 | |||
| XM_017003877.1 | XP_016859366.1 | |||
| AA401652.1 | ||||
| BQ269226.1 |
More Information
Set Membership: |
|

Genomic Map