Assay Details
Target Gene Details
Entrez Gene ID: | 105374811 |
Gene Name: | uncharacterized LOC105374811 |
Gene Aliases: |
- |
Location: |
Chr.2:75146352-75279143 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105374811 |
Target Gene Details
Entrez Gene ID: | 6869 |
Gene Name: | tachykinin receptor 1 |
Gene Aliases: |
NK1R, NKIR, SPR, TAC1R |
Location: |
Chr.2:75046463-75199519 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TACR1 | NM_001058.3 | NP_001049.1 | ||
| NM_015727.2 | NP_056542.1 | |||
| AK289765.1 | ||||
| AY462098.1 | AAR23925.1 | |||
| BC074911.2 | AAH74911.1 | |||
| BC074912.2 | AAH74912.1 | |||
| BC095528.1 | AAH95528.1 | |||
| M74290.1 | AAA60601.1 | |||
| M76675.1 | AAA59936.1 | |||
| M81797.1 | AAA59933.1 | |||
| M84425.1 | AAA36641.1 | |||
| M84426.1 | AAA36644.1 | |||
| S62045.1 | AAB20168.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759062 | Chr.2:75136297 - 75155584 on Build GRCh38 | Loss |
|
| nsv998145 | Chr.2:74969670 - 75241030 on Build GRCh38 | Loss |
|
| esv28202 | Chr.2:75153925 - 75155758 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map