Assay Details
Target Gene Details
Entrez Gene ID: | 4880 |
Gene Name: | natriuretic peptide C |
Gene Aliases: |
CNP, CNP2 |
Location: |
Chr.2:231922094-231926403 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPPC | NM_024409.3 | NP_077720.1 | ||
| XM_011511245.2 | 3 | 2392 | XP_011509547.1 | |
| DR003533.1 | ||||
| JF420840.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428408 | Chr.2:231672114 - 231968956 on Build GRCh38 | Gain+Loss |
|
| nsv508892 | Chr.2:231889469 - 231940618 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map