Assay Details
Target Gene Details
Entrez Gene ID: | 440944 |
Gene Name: | THUMPD3 antisense RNA 1 |
Gene Aliases: |
SETD5-AS1 |
Location: |
Chr.3:9385264-9397494 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THUMPD3-AS1 | NR_027007.3 | |||
| NR_132780.1 | ||||
| AK093640.1 | ||||
| AK128398.1 | BAC87421.1 | |||
| AL110136.1 | ||||
| BX953112.1 | ||||
| HY142091.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv277n21 | Chr.3:9390066 - 9453435 on Build GRCh38 | Loss |
|
| nsv821981 | Chr.3:9396551 - 9398182 on Build GRCh38 | Loss |
|

Genomic Map