Assay Details
Target Gene Details
Entrez Gene ID: | 101928142 |
Gene Name: | AADACL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:151751179-151928175 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AADACL2-AS1 | NR_110202.1 | |||
NR_110203.1 | ||||
DB454604.1 | ||||
HY022483.1 | ||||
HY205836.1 |
Target Gene Details
Entrez Gene ID: | 56670 |
Gene Name: | succinate receptor 1 |
Gene Aliases: |
GPR91 |
Location: |
Chr.3:151873643-151884619 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUCNR1 | NM_033050.5 | NP_149039.2 | ||
AF247785.1 | AAL95690.1 | |||
AK290420.1 | ||||
BC030948.1 | AAH30948.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829758 | Chr.3:151849745 - 152005470 on Build GRCh38 | Gain |
![]() ![]() |
esv2759189 | Chr.3:151757492 - 151945387 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
