Assay Details
Target Gene Details
Entrez Gene ID: | 93556 |
Gene Name: | EGF like and EMI domain containing 1, pseudogene |
Gene Aliases: |
C3orf50, NCRNA00259 |
Location: |
Chr.3:168249522-168830599 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGFEM1P | NR_021485.2 | |||
| BI461452.1 | ||||
| DR000794.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114825186,rs80028932] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map