Assay Details
Target Gene Details
Entrez Gene ID: | 389124 |
Gene Name: | IQ motif containing F5 |
Gene Aliases: |
- |
Location: |
Chr.3:51873721-51875584 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCF5 | NM_001145059.1 | NP_001138531.1 | ||
| BX106781.1 |
Target Gene Details
Entrez Gene ID: | 101928999 |
Gene Name: | IQCF5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:51873596-51875767 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCF5-AS1 | NR_109984.1 | 3 | 1593 | |
| AK097471.1 | 3 | 1593 | BAC05066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3817 | Chr.3:51843923 - 51877041 on Build GRCh38 | Insertion |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|

Genomic Map