Assay Details
Target Gene Details
Entrez Gene ID: | 257313 |
Gene Name: | urotensin 2B |
Gene Aliases: |
U2B, URP, UTS2D |
Location: |
Chr.3:191267155-191330536 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UTS2B | NM_198152.3 | NP_937795.2 | ||
| XM_011512631.1 | XP_011510933.1 | |||
| XM_017006091.1 | XP_016861580.1 | |||
| AB116021.1 | BAC98929.1 | |||
| AK090630.1 | ||||
| AY321313.1 | AAQ83883.1 | |||
| BC112166.1 | AAI12167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428428 | Chr.3:191219045 - 191497807 on Build GRCh38 | Gain |
|
| esv3599096 | Chr.3:191251629 - 191553437 on Build GRCh38 | Gain |
|
| esv2759209 | Chr.3:191149222 - 191561318 on Build GRCh38 | Gain+Loss |
|
| nsv997716 | Chr.3:191225917 - 191309710 on Build GRCh38 | Loss |
|
| esv3599094 | Chr.3:191225726 - 191310300 on Build GRCh38 | Loss |
|
| esv3599098 | Chr.3:191263515 - 191280402 on Build GRCh38 | Loss |
|
| nsv1012488 | Chr.3:191156997 - 191277390 on Build GRCh38 | Loss |
|
| nsv10369 | Chr.3:191276985 - 191293658 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map