Assay Details
Target Gene Details
Entrez Gene ID: | 254827 |
Gene Name: | N-acetylated alpha-linked acidic dipeptidase-like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:174439669-175809510 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
Target Gene Details
Entrez Gene ID: | 100874245 |
Gene Name: | NAALADL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:175773578-175776333 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAALADL2-AS1 | NR_046714.1 | |||
| AA905673.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526163 | Chr.3:175368755 - 176024960 on Build GRCh38 | Loss |
|
| esv3893821 | Chr.3:175553242 - 176177948 on Build GRCh38 | Loss |
|
| esv3569129 | Chr.3:175360717 - 176022379 on Build GRCh38 | Loss |
|
| nsv1001544 | Chr.3:175709801 - 175798369 on Build GRCh38 | Loss |
|
| esv2763715 | Chr.3:175360537 - 176029158 on Build GRCh38 | Loss |
|
| nsv1002662 | Chr.3:175655807 - 176170259 on Build GRCh38 | Gain |
|
| esv32881 | Chr.3:175372633 - 176002077 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map