Assay Details
Target Gene Details
Entrez Gene ID: | 13 |
Gene Name: | arylacetamide deacetylase |
Gene Aliases: |
CES5A1, DAC |
Location: |
Chr.3:151814008-151828488 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADAC | NM_001086.2 | NP_001077.2 | ||
| XM_005247104.4 | XP_005247161.1 | |||
| AK290628.1 | ||||
| AV705031.1 | ||||
| BC032309.1 | AAH32309.1 | |||
| L32179.1 | AAA35551.1 |
Target Gene Details
Entrez Gene ID: | 101928142 |
Gene Name: | AADACL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:151751179-151928175 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADACL2-AS1 | NR_110202.1 | |||
| NR_110203.1 | ||||
| DB454604.1 | ||||
| HY022483.1 | ||||
| HY205836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893786 | Chr.3:151796802 - 151828253 on Build GRCh38 | Loss |
|
| nsv592070 | Chr.3:151817634 - 151828253 on Build GRCh38 | Loss |
|
| nsv522472 | Chr.3:151743597 - 151864407 on Build GRCh38 | Loss |
|
| dgv1419e212 | Chr.3:151792302 - 151831226 on Build GRCh38 | Loss |
|
| esv2759189 | Chr.3:151757492 - 151945387 on Build GRCh38 | Loss |
|
| dgv4921n100 | Chr.3:151790048 - 151825513 on Build GRCh38 | Loss |
|
| esv25716 | Chr.3:151793701 - 151833212 on Build GRCh38 | Loss |
|
| dgv4920n100 | Chr.3:151784289 - 151830691 on Build GRCh38 | Loss |
|
| dgv8655n54 | Chr.3:151814106 - 151833114 on Build GRCh38 | Loss |
|
| esv3562727 | Chr.3:151792826 - 151836712 on Build GRCh38 | Deletion |
|
| esv2672544 | Chr.3:151793742 - 151833274 on Build GRCh38 | Deletion |
|
| nsv514175 | Chr.3:151793730 - 151832482 on Build GRCh38 | Loss |
|
| esv3598188 | Chr.3:151793743 - 151833279 on Build GRCh38 | Loss |
|
| dgv8652n54 | Chr.3:151792469 - 151833340 on Build GRCh38 | Loss |
|
| dgv4919n100 | Chr.3:151776103 - 151841876 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73166984] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map