Assay Details
Target Gene Details
Entrez Gene ID: | 11235 |
Gene Name: | programmed cell death 10 |
Gene Aliases: |
CCM3, TFAR15 |
Location: |
Chr.3:167683891-167735690 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDCD10 | NM_007217.3 | NP_009148.2 | ||
| NM_145859.1 | NP_665858.1 | |||
| NM_145860.1 | NP_665859.1 | |||
| XM_005247086.4 | XP_005247143.1 | |||
| XM_005247087.4 | XP_005247144.1 | |||
| XM_005247088.3 | XP_005247145.1 | |||
| XM_006713485.3 | XP_006713548.1 | |||
| XM_011512368.2 | XP_011510670.1 | |||
| XM_011512369.2 | XP_011510671.1 | |||
| XM_017005644.1 | XP_016861133.1 | |||
| XM_017005645.1 | XP_016861134.1 | |||
| AF022385.1 | AAB72225.1 | |||
| AK291130.1 | ||||
| BC002506.1 | AAH02506.1 | |||
| BC016353.1 | AAH16353.1 | |||
| CR457107.1 | CAG33388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv965231 | Chr.3:167693057 - 167694547 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map