Assay Details
Target Gene Details
Entrez Gene ID: | 100507086 |
Gene Name: | DLG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:197298247-197303751 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLG1-AS1 | NR_038289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599293 | Chr.3:196759100 - 197444213 on Build GRCh38 | Gain |
|
| nsv829830 | Chr.3:197161008 - 197354023 on Build GRCh38 | Gain |
|
| esv3893852 | Chr.3:197204950 - 197333165 on Build GRCh38 | Gain |
|
| nsv428431 | Chr.3:197128182 - 197316896 on Build GRCh38 | Gain+Loss |
|
| dgv5038n100 | Chr.3:197134594 - 197659420 on Build GRCh38 | Gain |
|
| nsv593116 | Chr.3:197183211 - 197617305 on Build GRCh38 | Gain |
|

Genomic Map