Assay Details
Target Gene Details
Entrez Gene ID: | 339874 |
Gene Name: | uncharacterized LOC339874 |
Gene Aliases: |
- |
Location: |
Chr.3:131325092-131381475 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC339874 | NR_038976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597785 | Chr.3:131350870 - 131415049 on Build GRCh38 | Gain |
|
| esv3597781 | Chr.3:131272356 - 131472358 on Build GRCh38 | Gain |
|
| nsv829724 | Chr.3:131366925 - 131557541 on Build GRCh38 | Gain |
|
| dgv934e214 | Chr.3:131361494 - 131382962 on Build GRCh38 | Loss |
|

Genomic Map