Assay Details
Target Gene Details
Entrez Gene ID: | 339829 |
Gene Name: | coiled-coil domain containing 39 |
Gene Aliases: |
CILD14, FAP59 |
Location: |
Chr.3:180614008-180679495 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC39 | NM_181426.1 | NP_852091.1 | ||
| AK304270.1 | ||||
| AL122120.1 | CAB59277.1 |
More Information
Set Membership: |
|

Genomic Map