Assay Details
Target Gene Details
Entrez Gene ID: | 4336 |
Gene Name: | myelin-associated oligodendrocyte basic protein |
Gene Aliases: |
- |
Location: |
Chr.3:39467573-39529497 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOBP | NM_001278322.1 | NP_001265251.1 | ||
| NM_182935.3 | NP_891980.1 | |||
| NR_003090.2 | ||||
| NR_103504.1 | ||||
| NR_103505.1 | ||||
| NR_103506.1 | ||||
| AK127034.1 | ||||
| AK290187.1 | ||||
| BC022471.1 | AAH22471.1 | |||
| BC075796.1 | AAH75796.1 | |||
| BC119694.1 | ||||
| BC119695.1 | ||||
| D28113.1 | BAA05659.1 | |||
| D28114.1 | BAA05660.1 | |||
| DA287397.1 | ||||
| DA343382.1 | ||||
| DA418624.1 | ||||
| EB387411.1 | ||||
| KU178193.1 | ||||
| KU178194.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759142 | Chr.3:39492781 - 39514006 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map