Assay Details
Target Gene Details
Entrez Gene ID: | 50650 |
Gene Name: | Rho guanine nucleotide exchange factor 3 |
Gene Aliases: |
GEF3, STA3, XPLN |
Location: |
Chr.3:56727418-57079308 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF3 | NM_001128615.1 | NP_001122087.1 | ||
| XM_005265186.4 | XP_005265243.1 | |||
| XM_005265187.3 | XP_005265244.1 | |||
| XM_011533764.1 | XP_011532066.1 | |||
| XM_011533765.1 | XP_011532067.1 | |||
| XM_011533766.2 | XP_011532068.1 | |||
| XM_017006502.1 | XP_016861991.1 | |||
| XM_017006503.1 | XP_016861992.1 | |||
| XM_017006504.1 | XP_016861993.1 | |||
| AF433662.1 | AAP97313.1 | |||
| AK302493.1 | ||||
| DC334772.1 |
Target Gene Details
Entrez Gene ID: | 100874200 |
Gene Name: | ARHGEF3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:56940040-56960854 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF3-AS1 | NR_046572.1 | |||
| BX090838.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590363 | Chr.3:56952459 - 56978210 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map