Assay Details
Target Gene Details
Entrez Gene ID: | 9718 |
Gene Name: | endothelin converting enzyme 2 |
Gene Aliases: |
- |
Location: |
Chr.3:184249657-184293031 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECE2 | NM_014693.3 | NP_055508.3 | ||
| NM_032331.3 | NP_115707.2 | |||
| AF428263.1 | AAL30386.1 | |||
| BC005835.2 | AAH05835.1 | |||
| BC012449.1 | AAH12449.1 | |||
| BC069005.1 | AAH69005.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528639 | Chr.3:184228486 - 184262628 on Build GRCh38 | Loss |
|
| nsv592666 | Chr.3:184220630 - 184288607 on Build GRCh38 | Loss |
|
| nsv829812 | Chr.3:184136655 - 184302302 on Build GRCh38 | Loss |
|
| nsv4146 | Chr.3:184252606 - 184285216 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map