Assay Details
Target Gene Details
Entrez Gene ID: | 100873945 |
Gene Name: | HLTF antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:149086332-149102823 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HLTF-AS1 | NR_046648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2726036 | Chr.3:149092060 - 149093654 on Build GRCh38 | Deletion |
|
| nsv829753 | Chr.3:148954528 - 149120758 on Build GRCh38 | Gain |
|
| nsv528849 | Chr.3:149054135 - 149117654 on Build GRCh38 | Gain |
|

Genomic Map