Assay Details
Target Gene Details
Entrez Gene ID: | 11170 |
Gene Name: | family with sequence similarity 107 member A |
Gene Aliases: |
DRR1, TU3A |
Location: |
Chr.3:58564112-58627610 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM107A | NM_001282714.1 | NP_001269643.1 | ||
| AK054720.1 | ||||
| DT216507.1 |
Target Gene Details
Entrez Gene ID: | 105377108 |
Gene Name: | FAM3D antisense RNA 1 |
Gene Aliases: |
lnc-KCTD6-3 |
Location: |
Chr.3:58607080-58634440 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM3D-AS1 | NR_134853.1 | |||
| BG215913.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4749n100 | Chr.3:58504269 - 60219035 on Build GRCh38 | Gain |
|
| esv3596253 | Chr.3:58621679 - 58677821 on Build GRCh38 | Gain |
|
| esv3893702 | Chr.3:58557785 - 58858605 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map