Assay Details
Target Gene Details
| Entrez Gene ID: | 114112 | 
| Gene Name: | thioredoxin reductase 3 | 
| Gene Aliases: | TGR, TR2, TRXR3 | 
| Location: | Chr.3:126607052-126655124 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 12 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| TXNRD3 | NM_001173513.1 | NP_001166984.1 | ||
| NM_052883.1 | NP_443115.1 | |||
| AF133519.1 | AAD39929.1 | |||
| AF171055.1 | AAD51325.1 | |||
| AK299502.1 | ||||
| BC030028.2 | AAH30028.1 | |||
| BC050032.1 | AAH50032.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv591524 | Chr.3:126614957 - 126669032 on Build GRCh38 | Gain |  TXNRD3  NUP210P1 | 
| dgv4852n100 | Chr.3:126607261 - 126669480 on Build GRCh38 | Gain |  TXNRD3NB  TXNRD3  NUP210P1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top