Assay Details
Target Gene Details
Entrez Gene ID: | 57501 |
Gene Name: | KIAA1257 |
Gene Aliases: |
- |
Location: |
Chr.3:128910186-129032431 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA1257 | NM_020741.2 | NP_065792.1 | ||
| XM_011513050.2 | XP_011511352.2 | |||
| XM_017006939.1 | XP_016862428.1 | |||
| XM_017006940.1 | XP_016862429.1 | |||
| XM_017006941.1 | XP_016862430.1 | |||
| XM_017006942.1 | XP_016862431.1 | |||
| XM_017006943.1 | XP_016862432.1 | |||
| XM_017006944.1 | XP_016862433.1 | |||
| XM_017006945.1 | XP_016862434.1 | |||
| XM_017006946.1 | XP_016862435.1 | |||
| XM_017006947.1 | XP_016862436.1 | |||
| XM_017006948.1 | XP_016862437.1 | |||
| XM_017006949.1 | XP_016862438.1 | |||
| XM_017006950.1 | XP_016862439.1 | |||
| XM_017006951.1 | XP_016862440.1 | |||
| XM_017006952.1 | XP_016862441.1 | |||
| XM_017006953.1 | XP_016862442.1 | |||
| AB033083.1 | ||||
| BC031632.1 | AAH31632.1 | |||
| BC038736.1 | AAH38736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3996 | Chr.3:128942534 - 128992631 on Build GRCh38 | Deletion |
|

Genomic Map