Assay Details
Target Gene Details
Entrez Gene ID: | 54965 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class X |
Gene Aliases: |
PIG-X |
Location: |
Chr.3:196712374-196736007 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGX | NM_001166304.1 | NP_001159776.1 | ||
| NM_017861.3 | NP_060331.3 | |||
| AA879458.1 | ||||
| AK000529.1 | BAA91233.1 | |||
| AK294340.1 | ||||
| BC022542.1 | AAH22542.1 | |||
| BG715890.1 | ||||
| CN404010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014526 | Chr.3:196645551 - 196733298 on Build GRCh38 | Gain |
|
| nsv829828 | Chr.3:196541250 - 196736770 on Build GRCh38 | Loss |
|
| nsv1009975 | Chr.3:196680410 - 196822871 on Build GRCh38 | Gain |
|
| nsv1001045 | Chr.3:196703082 - 196900384 on Build GRCh38 | Gain |
|
| nsv1003135 | Chr.3:196621850 - 197012082 on Build GRCh38 | Gain |
|
| nsv524832 | Chr.3:196727148 - 196735630 on Build GRCh38 | Loss |
|
| nsv1014692 | Chr.3:196515378 - 196804953 on Build GRCh38 | Gain |
|
| esv2763775 | Chr.3:196727069 - 197015419 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73072718] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map