Assay Details
Target Gene Details
Entrez Gene ID: | 23609 |
Gene Name: | makorin ring finger protein 2 |
Gene Aliases: |
HSPC070, RNF62 |
Location: |
Chr.3:12557014-12583713 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MKRN2 | NM_001271707.1 | NP_001258636.1 | ||
| NM_014160.4 | NP_054879.3 | |||
| AF302084.1 | AAG30426.1 | |||
| AK091318.1 | ||||
| AK298463.1 | ||||
| AL834512.1 | CAD39168.1 | |||
| BC001799.2 | AAH01799.2 | |||
| BC015715.1 | AAH15715.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000995 | Chr.3:12308425 - 12594756 on Build GRCh38 | Gain |
|
| esv3575548 | Chr.3:12521381 - 12631860 on Build GRCh38 | Gain |
|
| nsv1147269 | Chr.3:11914420 - 12871937 on Build GRCh38 | Deletion |
|
| nsv1112379 | Chr.3:11914395 - 12871920 on Build GRCh38 | Deletion |
|
| dgv850e199 | Chr.3:11914188 - 12872013 on Build GRCh38 | Deletion |
|
| esv2751977 | Chr.3:12452779 - 12887049 on Build GRCh38 | Gain |
|
| dgv763e201 | Chr.3:11909121 - 12871926 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113105671] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map