Assay Details
Target Gene Details
Entrez Gene ID: | 644990 |
Gene Name: | THRB antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:24494087-24500011 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THRB-AS1 | NR_046244.1 | |||
| AK130231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012975 | Chr.3:24459215 - 24574153 on Build GRCh38 | Gain |
|
| nsv999384 | Chr.3:24483854 - 24615382 on Build GRCh38 | Gain |
|
| dgv74n68 | Chr.3:24424371 - 24599102 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76215578] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map