Assay Details
Target Gene Details
Entrez Gene ID: | 152078 |
Gene Name: | PQ loop repeat containing 2 like |
Gene Aliases: |
C3orf55 |
Location: |
Chr.3:157543344-157601233 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PQLC2L | NM_001099777.3 | NP_001093247.1 | ||
| NM_001130001.2 | NP_001123473.1 | |||
| NM_001130002.2 | NP_001123474.1 | |||
| NM_001243729.1 | NP_001230658.1 | |||
| NR_024016.2 | ||||
| XM_017005754.1 | XP_016861243.1 | |||
| AA131625.1 | ||||
| BC029129.1 | AAH29129.1 | |||
| BC126187.1 | ||||
| BC126189.1 | ||||
| BE905784.1 | ||||
| BX113701.1 |
More Information
Set Membership: |
|

Genomic Map