Assay Details
Target Gene Details
Entrez Gene ID: | 101927777 |
Gene Name: | SATB1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:18445237-18530114 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SATB1-AS1 | NR_125803.1 | |||
| AW027730.1 | ||||
| BF115425.1 | ||||
| DR006704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589854 | Chr.3:18430796 - 18836070 on Build GRCh38 | Loss |
|

Genomic Map