Assay Details
Target Gene Details
Entrez Gene ID: | 114112 |
Gene Name: | thioredoxin reductase 3 |
Gene Aliases: |
TGR, TR2, TRXR3 |
Location: |
Chr.3:126607052-126655124 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNRD3 | NM_001173513.1 | 9 | 1265 | NP_001166984.1 |
| NM_052883.1 | 9 | 1265 | NP_443115.1 | |
| AF133519.1 | 9 | 933 | AAD39929.1 | |
| AF171055.1 | 9 | 939 | AAD51325.1 | |
| AK299502.1 | 9 | 1242 | ||
| BC030028.2 | 9 | 1243 | AAH30028.1 | |
| BC050032.1 | 9 | 1252 | AAH50032.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4852n100 | Chr.3:126607261 - 126669480 on Build GRCh38 | Gain |
|
| nsv460842 | Chr.3:126614957 - 126669032 on Build GRCh38 | Gain |
|
| nsv829718 | Chr.3:126629911 - 126809810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map