Assay Details
Target Gene Details
Entrez Gene ID: | 9881 |
Gene Name: | tetratricopeptide repeat and ankyrin repeat containing 1 |
Gene Aliases: |
LBA1 |
Location: |
Chr.3:36826817-36945662 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRANK1 | NM_014831.2 | NP_055646.2 | ||
| XM_006713432.3 | XP_006713495.1 | |||
| XM_011534289.2 | XP_011532591.1 | |||
| XM_017007570.1 | XP_016863059.1 | |||
| XM_017007571.1 | XP_016863060.1 | |||
| XM_017007572.1 | XP_016863061.1 | |||
| XM_017007573.1 | XP_016863062.1 | |||
| AB002340.3 | BAA20800.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834661 | Chr.3:36759557 - 36951666 on Build GRCh38 | Loss |
|
| nsv834662 | Chr.3:36830593 - 37033470 on Build GRCh38 | Loss |
|
| nsv998118 | Chr.3:36819990 - 36848132 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map