Assay Details
Target Gene Details
Entrez Gene ID: | 51208 |
Gene Name: | claudin 18 |
Gene Aliases: |
SFTA5, SFTPJ |
Location: |
Chr.3:137998816-138033652 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLDN18 | NM_001002026.2 | NP_001002026.1 | ||
| NM_016369.3 | NP_057453.1 | |||
| AF221069.1 | AAF26448.1 | |||
| AF349452.1 | AAL15637.1 | |||
| AK098474.1 | ||||
| AK298103.1 | ||||
| AY358479.1 | AAQ88843.1 | |||
| BC142708.1 | ||||
| BC146668.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
| nsv460863 | Chr.3:137964915 - 138022297 on Build GRCh38 | Loss |
|
| esv2751978 | Chr.3:137996460 - 138171908 on Build GRCh38 | Loss |
|
| esv2759180 | Chr.3:137892947 - 138214993 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114302616] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map