Assay Details
Target Gene Details
Entrez Gene ID: | 254827 |
Gene Name: | N-acetylated alpha-linked acidic dipeptidase-like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:174439669-175809510 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAALADL2 | XM_006713560.2 | XP_006713623.1 | ||
| XM_017006075.1 | XP_016861564.1 | |||
| XM_017006077.1 | XP_016861566.1 | |||
| XM_017006078.1 | XP_016861567.1 | |||
| XM_017006080.1 | XP_016861569.1 | |||
| XM_017006082.1 | XP_016861571.1 | |||
| AJ607398.1 | ||||
| AJ607399.1 | ||||
| AJ607401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005730 | Chr.3:174266156 - 174829055 on Build GRCh38 | Gain |
|
| nsv4124 | Chr.3:174461292 - 174478318 on Build GRCh38 | Deletion |
|
| nsv520908 | Chr.3:174083961 - 174816144 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73881169] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map