Assay Details
Target Gene Details
Entrez Gene ID: | 152137 |
Gene Name: | coiled-coil domain containing 50 |
Gene Aliases: |
C3orf6, DFNA44, YMER |
Location: |
Chr.3:191329082-191398670 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC50 | NM_174908.3 | NP_777568.1 | ||
| NM_178335.2 | NP_848018.1 | |||
| XM_011512460.1 | XP_011510762.1 | |||
| AJ416916.2 | CAC95196.1 | |||
| AJ557013.1 | CAD89526.1 | |||
| BC065004.1 | AAH65004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428428 | Chr.3:191219045 - 191497807 on Build GRCh38 | Gain |
|
| esv3599096 | Chr.3:191251629 - 191553437 on Build GRCh38 | Gain |
|
| esv2759209 | Chr.3:191149222 - 191561318 on Build GRCh38 | Gain+Loss |
|
| esv3599102 | Chr.3:191304692 - 191335341 on Build GRCh38 | Loss |
|
| nsv4169 | Chr.3:191318509 - 191345676 on Build GRCh38 | Deletion |
|
| nsv998208 | Chr.3:191303645 - 191457561 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map