Assay Details
Target Gene Details
Entrez Gene ID: | 79825 |
Gene Name: | EF-hand and coiled-coil domain containing 1 |
Gene Aliases: |
C3orf73, CCDC48 |
Location: |
Chr.3:129001604-129040742 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCC1 | NM_024768.2 | NP_079044.2 | ||
| XM_005247781.3 | XP_005247838.1 | |||
| XM_011513161.2 | XP_011511463.1 | |||
| XM_011513163.2 | XP_011511465.1 | |||
| XM_011513164.2 | XP_011511466.1 |
Target Gene Details
Entrez Gene ID: | 57501 |
Gene Name: | KIAA1257 |
Gene Aliases: |
- |
Location: |
Chr.3:128910186-129032431 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA1257 | XM_011513050.2 | XP_011511352.2 | ||
| XM_017006939.1 | XP_016862428.1 | |||
| XM_017006940.1 | XP_016862429.1 | |||
| XM_017006941.1 | XP_016862430.1 | |||
| XM_017006942.1 | XP_016862431.1 | |||
| XM_017006943.1 | XP_016862432.1 | |||
| XM_017006945.1 | XP_016862434.1 | |||
| XM_017006946.1 | XP_016862435.1 | |||
| XM_017006947.1 | XP_016862436.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591650 | Chr.3:128986168 - 129177231 on Build GRCh38 | Loss |
|
| nsv997717 | Chr.3:129006263 - 129081989 on Build GRCh38 | Loss |
|
| nsv519919 | Chr.3:128993535 - 129022799 on Build GRCh38 | Loss |
|
| esv3597731 | Chr.3:128990006 - 129023340 on Build GRCh38 | Gain |
|
| esv3597730 | Chr.3:128986215 - 129087514 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112531856] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map