Assay Details
Target Gene Details
Entrez Gene ID: | 5276 |
Gene Name: | serpin family I member 2 |
Gene Aliases: |
MEPI, PANCPIN, PI14, TSA2004 |
Location: |
Chr.3:167441789-167474132 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINI2 | NM_001012303.2 | NP_001012303.1 | ||
| NM_006217.4 | NP_006208.1 | |||
| AB006423.1 | BAA33766.1 | |||
| AF130470.1 | AAD34723.1 | |||
| AK226025.1 | ||||
| AK293391.1 | ||||
| BC027859.2 | AAH27859.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv592495 | Chr.3:167077165 - 167655374 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148932798] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map