Assay Details
Target Gene Details
Entrez Gene ID: | 646424 |
Gene Name: | serine peptidase inhibitor, Kazal type 8 (putative) |
Gene Aliases: |
- |
Location: |
Chr.3:48306846-48330311 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINK8 | XM_017007046.1 | XP_016862535.1 | ||
| XM_017007047.1 | XP_016862536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834683 | Chr.3:48260014 - 48451855 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map