Assay Details
Target Gene Details
Entrez Gene ID: | 344967 |
Gene Name: | acyl-CoA thioesterase 7 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.4:40042917-40057199 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC344967 | NR_027277.2 | |||
| AK125299.1 | BAC86118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5194n100 | Chr.4:39916983 - 40119942 on Build GRCh38 | Gain |
|
| nsv594033 | Chr.4:39934245 - 40095937 on Build GRCh38 | Gain |
|
| nsv594044 | Chr.4:39983318 - 40185488 on Build GRCh38 | Gain |
|
| nsv594032 | Chr.4:39893368 - 40131994 on Build GRCh38 | Gain |
|
| nsv594043 | Chr.4:39977946 - 40049444 on Build GRCh38 | Gain |
|
| esv2422397 | Chr.4:39826035 - 40047821 on Build GRCh38 | Duplication |
|
| esv3575688 | Chr.4:39922679 - 40113190 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map