Assay Details
Target Gene Details
Entrez Gene ID: | 100144602 |
Gene Name: | EPHA5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:65669961-65693386 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA5-AS1 | NR_034138.1 | |||
| BC017721.1 | AAH17721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422517 | Chr.4:65582828 - 65694176 on Build GRCh38 | Duplication |
|
| nsv1014463 | Chr.4:65294286 - 65989398 on Build GRCh38 | Gain |
|
| nsv291906 | Chr.4:65672448 - 65676968 on Build GRCh38 | Deletion |
|

Genomic Map